missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Dextrose Tryptone Agar
For cultivation of thermophilic flat-sour microorganisms associated with food spoilage
Brand: BD 280100
This item is not returnable.
View return policy
Description
Specifications
COMPF | |
Dehydrated | |
Agar | |
Cultivation of thermophilic “flat-sour” microorganisms associated with food spoilage | |
500 g |
Refrigerate | |
Dextrose Tryptone Agar | |
Tryptone 8g/L, Peptone 2g/L, Dextrose 5g/L, Agar 15g/L, Bromcresol Purple 0.04g/L | |
Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: Dextrose Tryptone Agar > Dextrose Tryptone Agar; 500g
Spot an opportunity for improvement?Share a Content Correction