missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Phenylalanine Agar
Click to view available options
:
Phenylalanine Agar; 500g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Certifications/Compliance | BAM |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Phenylalanine Agar |
| Product Type | Agar |
| Formulation | DL-Phenylalanine 2g/L, Yeast extract 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 1g/L, Agar 12g/L |
| For Use With (Application) | For differentiation of enteric Bacilli on the basis of their ability to produce Phenylpyruvic Acid by oxidative deamination |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction