Learn More
BD Difco™ Dehydrated Culture Media: Rose Bengal Agar Base
Click to view available options
:
Rose Bengal Agar Base; 500g
Description
- Formulation: Soy Peptone 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Rose Bengal 0.05g/L, Agar 15g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
SMWW
Specifications
Specifications
| Certifications/Compliance | SMWW |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Rose Bengal Agar Base |
| Product Type | Agar |
| Formulation | Soy Peptone 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Rose Bengal 0.05g/L, Agar 15g/L |
| For Use With (Application) | Rose Bengal antimicrobic supplement C in isolating and enumerating Yeasts and molds |
| Packaging Type | Bottle |
| Quantity | 500 g |