Learn More
Bel-Art™ SP Scienceware™ Microtube Cutter
Product Code. p-200479 Shop All SP Scienceware Products
Click to view available options
Material:
Chrome-plated Steel
Steel
Product Type:
Microtube Cutter
Replacement Blade
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 11730635
Brand: Bel-Art™ F210180000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Safely, accurately and easily sections microcentrifuge tubes and eliminates the hazards of using razor blades or knives
  • Makes it easy to retrieve radioactive pellets from the tips of tubes without resuspension or the risk of splashed radioactive substances
  • Works well with polyethylene c or polypropylene microcentrifuge tubes up to 1.5mL
  • Chrome-plated body with replaceable steel blade
  • Maximum cut diameter is 9.4mm (0.370 in.)
Not recommended for polystyrene tubes.
Specifications
Material Chrome-plated Steel
For Use With Microtube cutter
Product Type Microtube Cutter