Learn More
Bel-Art™ Spinbox™ Spinplus™ Magnetic Stirring Bar Assortment
Shop All SP Scienceware ProductsDescription
Includes
Octagon figured 16 assorted bars, standard and micro; standard bars with pivot ring Octagon shaped 12 assorted bars, standard bars with pivot rings Round shaped 22 assorted bars with removable pivot rings on bars longer than 1 in. Metric Polygon shaped 18 assorted bars Color-coded, octagon shaped 24 assorted bars with pivot ring Color-coded micro bars 12 assorted bars Egg shaped bars 13 assorted bars Spinplus in. + shaped bars 5 assorted bars
- Convenient storage box segregates sizes and styles for easy, no-rummage organization
- FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Available in octagon, egg and Spinplus™ shapes
- Autoclavable
Certifications
ISO 9001:2008
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | FDA, USP Class VI, ISO 9001:2008, SGS |
| Includes | 16 assorted standard and micro octagonal bars; 12 assorted standard octagonal bars with pivot ring; 22 assorted standard round bars with pivot ring (removable pivot ring on bars longer than 1 in.); 18 assorted metric polygon colour-coded bars; 24 assorted colour-coded micro bars with pivot ring; 12 assorted egg shaped bars; 13 assorted Spinplus including 5 '+' shaped bars |
| Coating | PTFE, FDA-grade, USP Class VI |
| Diameter (Metric) | 9.7 mm, 12.7 mm, 16 mm, 19 mm |
| Length (Metric) | 19 mm, 25.4 mm, 31.8 mm, 38.1 mm, 41.4 mm, 50.8 mm, 76.2 mm |
| Material | Alnico Magnet |
| Quantity | 13 |
| Shape | Egg |
| Type | Magnetic Stir Bar Set |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.