missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEX1/2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12390-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects both BEX1 and BEX2.
BEX1/BEX2 belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. BEX1/BEX2 has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| BEX1/2 | |
| Polyclonal | |
| Unconjugated | |
| BEX1 | |
| BEX1, BEX1/2, BEX2, brain expressed X linked 2, hBex2, Protein BEX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12069, 19716, 363498, 501625, 55859, 84707 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3MKQ1, Q3MKQ2, Q9BXY8, Q9HBH7, Q9R224, Q9WTZ8 | |
| Bex1, Bex2 | |
| BEX1/2 Fusion Protein Ag3066 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction