Learn More
Krüss Optronic™ Binocular Microscope
Brand: Krüss Optronic™ MBL2000
This item is not returnable.
View return policy
Specifications
Double lens ABBE Condenser, NA 1.25, with centering, height adjustment and filter holder | |
Inclined optical head | |
Achromatic: 4X 0.10 NA 17.04 mm working distance, 10X 0.25 NA 8.05 mm working distance, 40X 0.65 NA 0.32 mm working distance, 100X 1.25 NA 0.13 mm working distance |
10X/25 mm | |
Halogen |