Learn More
Tocris Bioscience™ BIO 5192
Highly potent and selective inhibitor of integrin α4β1
Brand: Tocris Bioscience™ 5051/50
1169.73 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
BIO 5192 is a highly selective and potent inhibitor of integrin α4β1 (Very Late Antigen-4; VLA-4) (Kd < 10 pM). Selectively binds α4β1 over a range of other integrins (IC50 values are 1.8, 138, 1053, > 500 and > 10,000 nM for α4β1, α9β1, α2β1, α4β7 and αIIbβ3, respectively). Induces a 30-fold increase in mobilization of murine hematopoietic stem and progenitor cells; displays a 3-fold additive effect with AMD 3100 (Cat. No. 3299). Decreases oxidative damage and improves neurological function following spinal cord injury in rats.Specifications
>98% | |
BIO 5192 | |
BIO 5192 |
50 mg | |
817.78 |
For Research Use Only