missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Biogen™ Triple Baffled Erlenmeyer Flask

Provide improved agitation and gas transfer with these triple-baffled flasks. Witeg™ Biogen™ Triple Baffled Erlenmeyer Flask is designed with three internal indents for greater mixing and long, straight necks that accommodate metal caps and reduce splashing.

Brand:  Witeg™ 5 507 107

 View more versions of this product

Product Code. 10694582

  • 82.25 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Graduated.
  • Labelling space.
  • Triple baffled.
  • Long neck.
Specifications

Specifications

Metal
38 mm
Orbital shaker
Long
Yes
Cap
Triple-baffled
1000 mL
232 mm
Erlenmeyer
Yes
131 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions