Learn More
Thermo Scientific BIOS adapter 42 x 10mL blood tubes Set of 2
Brand:  Thermo Scientific 75003342
BIOS adapter 42 x 10mL blood tubes Set of 2
Product Code. 17800695
  • ONLINE EXCLUSIVE PRICE
    960.00 €
    908.00 € / Each
    Save 52.00 €
    5% Off