missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation BioSuite DEAE Anion-Exchange Column
Click to view available options
Length (Metric):
75 mm
150 mm
Unit Size:
Each
Description
- The availability of four separation chemistries provides chromatographers with the flexibility required to develop methods that separate proteins and / or peptides based upon minor charge differences.
- Nonporous (NP) and porous IEX columns are also available. Superior chromatographic resolution is possible using the nonporous IEX offerings however, greater binding capacity is obtained with the porous selections.
- In addition, selected BioSuite ion-exchange columns are available in PEEK hardware as well as in 21.5 mm preparative column sizes.
Specifications
Specifications
| Diameter (Metric) | 7.5 mm |
| Length (Metric) | 75 mm |
| Particle Shape | Spherical |
| Particle Size | 10 μm |
| Pore Size | 1000 Å |
| Quantity | 1/Ea. |
| USP Type | L23 |
| Type | Column |
| For Use With (Application) | Protein |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction