missing translation for 'onlineSavingsMsg'
Learn More
Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals Product Code.: 11323946

Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals

Product Code. 11323946
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11323946

Brand: Thermo Scientific Alfa Aesar L17055.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6334-96-9
Molecular Formula C8H16Cl2O
Molecular Weight (g/mol) 199.115
MDL Number MFCD00019006
InChI Key PVBMXMKIKMJQRK-UHFFFAOYSA-N
Synonym bis 4-chlorobutyl ether, 4,4'-dichlorodibutyl ether, ether, bis 4-chlorobutyl, 4-chlorobutyl ether, di chlorobutyl ether, butane, 1,1'-oxybis 4-chloro, 1,1'-oxydi-4-chlorobutane, 1,1'-oxybis 4-chlorobutane
PubChem CID 22800
IUPAC Name 1-chloro-4-(4-chlorobutoxy)butane
SMILES C(CCCl)COCCCCCl

Specifications

Density 1.081
Boiling Point 129°C to 131°C (10 mmHg)
Linear Formula Cl(CH2)4O(CH2)4)Cl
Refractive Index 1.458
Quantity 25 g
Beilstein 1739861
Formula Weight 199.12
Percent Purity 99%
Chemical Name or Material Bis(4-chlorobutyl) ether

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10159885 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberL17055.06,L17055.22,L17055.14
Lot Number10159836 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberL17055.06,L17055.22,L17055.14
Lot Number10159885 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL17055.06,L17055.22,L17055.14
Lot Number10159836 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL17055.06,L17055.22,L17055.14
Lot NumberL05S035 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL17055.06,L17055.22,L17055.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.