missing translation for 'onlineSavingsMsg'
Learn More

Black Fine Nib Marker Pens

Product Code. 10058080
Encompass_Preferred
Click to view available options
:
Black
This item is not returnable. View return policy

Product Code. 10058080

Brand: Aspen surgical 140020PDC

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this pen for a wide range of labeling applications. Black Fine Nib Marker Pens are particularly suitable for tissue and cell culture applications.

  • With hard wearing, fine fiber point
  • For labeling most materials (plastic, metal, china, glass etc.)
  • The non-toxic ink is dye, bleach and wash-resistant and also resists solvents including alcohol and xylene

Specifications

Color Black
For Use With (Application) For marking of Plastics, Metal, Porcelain, Glass, tissue and cell cultures
Quantity 5/Pk.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.