Learn More
Brady™ BMP™61 Series Printer Ribbon - R6000 Resin, Black
BMP™61 Series ribbon cartridges allow users to change ribbons in seconds.
Brand:  Brady™ 148128
Product Code. 16359141
  • 38.85 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
  • One-color ribbon for printing a single color onto a label
  • R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Automatic label formatting with smart cell technology and easy loading
Specifications
Specifications
Black