missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Alanine, 99+%, Thermo Scientific Chemicals

34.00 € - 93.30 €

Chemical Identifiers

CAS 15761-38-3
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037225
InChI Key QVHJQCGUWFKTSE-YFKPBYRVSA-N
Synonym boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
PubChem CID 85082
SMILES C[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10571111
View Documents Promotion Details
Thermo Scientific Acros
275490100
10 g Glass Bottle
34.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10229263
View Documents Promotion Details
Thermo Scientific Acros
275490500
50 g Glass Bottle
93.30 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15761-38-3
189.21
QVHJQCGUWFKTSE-YFKPBYRVSA-N
85082
C8H15NO4
MFCD00037225
boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

81.0°C to 84.0°C
White
C8H15NO4
(CH3)3COC(O)NHC(CH3)COOH
04, 2507
QVHJQCGUWFKTSE-YFKPBYRVSA-N
(2S)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
189.21
189.21
Granular Powder
15761-38-3
99+%
Authentic
MFCD00037225
boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
C[C@H](NC(=O)OC(C)(C)C)C(O)=O
− 26.00
85082
≥99%
BOC-L-Alanine
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only