Learn More
Bohlender™ BOLA™ Spare O-rings for Stopcocks
Click to view available options
Material:
NBR
Unit Size:
Pack of 50
Description
- O-rings made of NBR
- For BOLA™ Eco Stopcock Plugs
- Taper: 1:10
- Thread M: 7 mm
- Suitable for size: NS 10
Specifications
Specifications
| Type | Spare O-ring |
| Material | NBR |
| For Use With (Equipment) | For BOLA™ Eco Stopcock Plugs |