Learn More
Bohlender™ BOLA™ Tri-clamp Gaskets
Double-sided tri-clamp gaskets with elastic PTFE sleeve and silicone inlay.
Brand:  Bohlender™ D930-44
 View more versions of this product
Product Code. 17137344
  • 39.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Silicone ring with double-sided, elastic washer made of PTFE
  • Suitable for Tri-clamp connections as per DIN 32 676
  • Universal chemical resistance
  • Product is only exposed to PTFE
Specifications
Specifications
Tubing with 32 mm I.D.
2 in.
PTFE, Silicone
DN 32
50.5 mm