Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
Brand: BRAND™ 144630
7.19 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Hollow
- Hexagonal grip
Specifications
1/Pk. | |
NS 24/29 | |
Ground Joint Stopper |
NS 24/29 | |
Borosilicate Glass | |
Conical |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.