Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04004128
 View more versions of this product
Product Code. 10748921
  • 79.00 € / Pack of 2
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Without scale
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
50 mL
Borosilicate Glass
2Pack
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug > Capacity: 50mL