Learn More
Fisherbrand™ General Purpose Borosilicate Homogenizer Mortars
For use during the initial stages of sample preparation to break down and disrupt a range of materials such as animal or plant tissues, foodstuffs and soil. Fisherbrand™ General Purpose Borosilicate Homogenizer Mortars are tissue grinders manufactured from durable borosilicate glass.
Brand: Fisherbrand™ 22004F
Description
- Autoclavable
- Available in a range of capacities
- For use with Fisherbrand Borosilicate Homogenizer Pestles
Specifications
0.152 - 0.254mm | |
30 mL | |
Homogenizers | |
250mm |
Autoclavable | |
Borosilicate Glass | |
105mm | |
Glass |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ General Purpose Borosilicate Homogenizer Mortars > Capacity: 30mL
Spot an opportunity for improvement?Share a Content Correction