missing translation for 'onlineSavingsMsg'
Learn More

Kimble Chase™ Borosilicate Glass Shell Vials

Product Code. 10123752
Click to view available options
Capacity (Metric):
2 mL
16 mL
Unit Size:
Pack of 1440
Pack of 576
This item is not returnable. View return policy

Product Code. 10123752

Brand: Kimble Chase™ 609314

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Kimble™ Borosilicate Glass Shell Vials for storage of dry contents.

  • Straight-sided
  • Clear N-51A borosilicate glass
  • Without caps
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 16 mL
Material Borosilicate Glass
For Use With (Application) Ideal for Storing Dry Products
Height (Metric) 70 mm
Type Shell Vial
Graduated Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.