Learn More
Reitenspiess™ Bottle Brush
Clean, disinfect and sterilize your laboratory bottles using the Reitenspiess™ Bottle Brush. With a range of lengths and bristle types, the brush can be specified to your specific glassware needs.
Brand:  Reitenspiess™ 21400234
 View more versions of this product
Product Code. 10349503
  • 12.20 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Variety of bristle types
  • 1 each in package
  • Variety of lengths and diameters
Specifications
Specifications
Bottle Brush
40 mm
1 Ea.
Natural
390 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Reitenspiess™ Bottle Brush > Diameter (metric) Outer: 40mm