Learn More
Lonza™ Bovine Pituitary Extract (BPE), 13mg/mL, 2 mL
Useful supplement in the culture of various epithelial cell types.
Brand:  Lonza™ CC-4009
Product Code. 11685480
  • 420.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Suitable for use in the culture of various epithelial cell types.
  • The product is included in SingleQuots™ Kits associated with various media systems for epithelial cell types.
  • Available at a concentration of 13 mg/mL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13 mg/mL
Bovine Pituitary Extract
2 mL