Learn More
3M™ Bowie and Dick Steritest
3M™ Bowie and Dick Steritest is suitable for use with autoclaves.
Brand: 3M™ 7019941
This item is not returnable.
View return policy
Description
- Class 2 indicator according to EN ISO 11140-1
- Non-reflective surface of indicator sheet for better visual assessment of color change
- Easy to use
- Conformance to standards
- Dimensions: 3 x 13 x 13 cm
Specifications
Class 2, EN ISO 11140-1 | |
Autoclaves | |
20 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Bowie and Dick Steritest > For Use With: Autoclaves
Spot an opportunity for improvement?Share a Content Correction