Learn More
JDM SCIENTIFIC Box Scientific Heliport
Expanded thawing surface for large sample arrays
Brand: JDM SCIENTIFIC 1-300-70000
Alert:
ATTENTION: US power plug - separate EU adapter required (not included)
Description
- Thaws frozen samples in a fraction of the time using only ambient air
- Accommodates 11 microplates and most standard tubes.
Specifications
Thawing Unit | |
13 cm | |
19 cm |
11 Microplates and Most Standard Tubes | |
76.5 cm | |
12 VDC/1.4 A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.