Learn More
Trajan™ BP1 GC Capillary Columns
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Unit Size:
Each
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
| Max. Temperature | 300/320°C |
| Functionality/Interaction Type | Van der Waals interaction |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | 100% Dimethyl Polysiloxane |
| USP Type | G1, G2, G38 |
| For Use With (Application) | Phenols Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines |
| Temperature Range | -60°C to +300/320°C |
| Film Thickness | 3 μm |
| Length (Metric) | 60 m |
| Packing Material | Forte™ BP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu