Learn More
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Service kits for VACUUBRAND NT series Chemistry design pumps.
Brand: VACUUBRAND™ 20696870
358.15 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sets include appropriate diaphragms, valves and sealing rings (if applicable)
- Wrench not included.
Specifications
MD 4C NT VARIO, PC3003 VARIO, PC3004 VARIO | |
Service Kit |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
13Hours
10Minutes
9Seconds
38New Offer Save Up to 92%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems > For MD4C NT VARIO, PC3003 VARIO, PC3004 VARIO
Spot an opportunity for improvement?Share a Content Correction