Learn More
Branson Ultrasonics™ Microtips for Sonifiers SLPe, SLPt, 150E and 150T
Product Code. 11715978
Click to view available options
Volume Range (Metric):
3 mL or less
3 to 10 mL
5 to 25 mL
Diameter (Metric):
0.22 cm
0.32 cm
0.48 cm
Unit Size:
Each
Product Code. 11715978
Brand: Branson Ultrasonics™ 1091221066
Please to purchase this item. Need a web account? Register with us today!
Designed to process samples to the smallest diameter vessels.
  • Can handle volumes up to 100mL
  • Working length: 4 in.
  • Sound proof enclosure
Order Info
0.125 in. microtip is provided with the 150 watt units
Specifications
Product Type SLP Microtip
Capacity (Metric) 1 mL
Volume Range (Metric) 3 mL or less
Diameter (English) 0.09 in.
Diameter (Metric) 0.22 cm
Length (English) 4 in.
Length (Metric) 10.16 cm
For Use With (Application) Ideal for processing samples to the smallest diameter vessels
For Use With (Equipment) SLPe, SLPt, 150E and 150T Sonifier™