missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BRAP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559145

Product Code. 15707011

  • 572.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SLVVIRLELA EHSPVPAGFG FSAAAGEMSD EEIKKTTLAS AVACLEGKSP GEKVAIIHQH LGRREMTDVI IETMKSNPDE LKTTVEERKS SEA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

Negatively regulates MAP kinase activation by limiting the formation of Raf/MEK complexes probably by inactivation of the KSR1 scaffold protein. Also acts as a Ras responsive E3 ubiquitin ligase that, on activation of Ras, is modified by auto-polyubiquitination resulting in the release of inhibition of Raf/MEK complex formation. May also act as a cytoplasmic retention protein with a role in regulating nuclear transport.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BRAP
Polyclonal
Unconjugated
BRAP
3010002G07Rik; BRAP; BRAP2; BRCA1 associated protein; BRCA1-associated protein; galectin-2-binding protein; IMP; Impedes mitogenic signal propagation; renal carcinoma antigen NY-REN-63; RING finger protein 52; RING-type E3 ubiquitin transferase BRAP2; RNF52
Rabbit
Antigen affinity chromatography
RUO
687346, 72399, 8315
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7Z569, Q99MP8
BRAP
Recombinant protein corresponding to Human BRAP. Recombinant protein control fragment (Product #RP-97663).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.