missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BRD3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand: Abnova™ H00008019-T01.100uL
This item is not returnable.
View return policy
Specifications
293T | |
BRD3 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 61.27kDa | |
Transient overexpression cell lysate was tested with anti-BRD3 antibody (Mfr. No. H00008019-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
BC032124.2 | |
pCMV-BRD3 full-length | |
100 μL | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction