Learn More
BRD9539, MedChemExpress
MedChemExpress BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1.
Brand: Medchem Express HY-15647
Description
BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1.Specifications
BRD9539 | |
White | |
C24H21N3O3 | |
DMSO : 16.67 mg/mL (41.73 mM; Need ultrasonic) | |
399.44 | |
99.2% | |
Solid |
1374601-41-8 | |
Research | |
25 mg | |
O=C(C1=CC=C2C(N=C(NC(C3=CC=CC=C3)=O)N2CCCC4=CC=CC=C4)=C1)O | |
399.44 | |
Research | |
Cancer-Kinase/protease |