Learn More
BRD9539, MedChemExpress
MedChemExpress BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1.
Brand:  Medchem Express HY-15647
Encompass_Preferred
 View more versions of this product
Product Code. 16443486
  • POA / 25mg
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1.
Specifications
Specifications
BRD9539
White
C24H21N3O3
DMSO : 16.67 mg/mL (41.73 mM; Need ultrasonic)
399.44
99.2%
Solid
1374601-41-8
Research
25 mg
O=C(C1=CC=C2C(N=C(NC(C3=CC=CC=C3)=O)N2CCCC4=CC=CC=C4)=C1)O
399.44
Research
Cancer-Kinase/protease
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BRD9539, MedChemExpress