missing translation for 'onlineSavingsMsg'
Learn More

Brefeldin A, 98%, Thermo Scientific Chemicals

Product Code. 15474739
Click to view available options
Quantity:
10 mg
25 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

Solubility
Soluble in dimethylsulfoxide, dichloromethane and ethanol. Slightly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4
Molecular Weight (g/mol) 280.36
MDL Number MFCD00083258,MFCD12913297
InChI Key KQNZDYYTLMIZCT-PNFJWZTBSA-N
Synonym brefeldin a, cyanein, decumbin, bfa
PubChem CID 133640201
SMILES C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

Specifications

Melting Point 201°C to 205°C
Color White
Quantity 25 mg
UN Number UN2811
Beilstein 25191
Merck Index 14,1369
Solubility Information Soluble in dimethylsulfoxide,dichloromethane and ethanol. Slightly soluble in water.
IUPAC Name (1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one
Formula Weight 280.36
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brefeldin A
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.