missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocriptine mesylate, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Bromocriptine mesylate is a selective D3 and D2 dopamine receptor agonist (Ki values are ∼ 5 nM and ∼ 8 nM respectively). Activates both isoforms of D2 receptor (EC50 values are 0.10-0.12 nM and 4.4 nM for D2L and D2S receptors, respectively. Bromocriptine shows anti-Aβ effects that are increased in combination with Cromolyn (Cat. No. 6424) and Topiramate (Cat. No. 3620). Also lowers glucose and increases insulin sensitivity in vivo.
Specifications
Specifications
CAS | 22260-51-1 |
Molecular Formula | C33H44BrN5O8S |
Quantity | 50 mg |
Synonym | bromocriptine mesylate |
InChI Key | NOJMTMIRQRDZMT-GPLFXVSLSA-N |
SMILES | CC(C)CC1C(=O)N2CCCC2C3(N1C(=O)C(O3)(C(C)C)NC(=O)C4CN(C5CC6=C(NC7=CC=CC(=C67)C5=C4)Br)C)O.CS(=O)(=O)O |
Molecular Weight (g/mol) | 750.706 |
PubChem CID | 126968774 |
Formula Weight | 750.7 |
Percent Purity | >98% |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction