missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BEGM™ Bronchial Epithelial SingleQuots™ Kit

Contains growth factors and supplements required for culturing Bronchial Epithelial cells (basal media not included)

Brand:  Lonza™ CC-4175


Product Code. 11675500

  • 330.00 € / 506mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 1 x Orange Cap Vial with BPE 2.00 mL
  • 1 x Lilac Cap Vial with Insulin 0.50 mL
  • 1 x Natural Cap Vial with Hydrocortisone 0.50 mL
  • 1 x Red Cap Vial with GA-1000 0.50 mL
  • 1 x Amber Vial with Retinoic Acid 0.50 mL
  • 1 x Natural Cap Vial with Transferrin 0.50 mL
  • 1 x Amber Vial with Triiodothyronine 0.50 mL
  • 1 x Amber Vial with Epinephrine 0.50 mL
  • 1 x Green Cap Vial with hEGF 0.50 mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SingleQuots™ Kit
1 Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BEGM SingleQuots

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Promotions

Promotions