Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Product Code. 10761041
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
150 mL
200 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
7/16 NS Standard Ground
Unit Size:
Each
Pack of 2
Product Code. 10761041
Brand: Hirschmann™ 2640294
Please to purchase this item. Need a web account? Register with us today!
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Color Brown
Closure Material Glass
Color Brown
Capacity (Metric) 2000 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 370 mm
Limit of Error ±0.6 mL
Standard Taper Joint 29/32 NS Standard Ground
Tolerance ±0.6 mL
Graduated Yes
Class A