Learn More
BTN1A1 Monoclonal Antibody (OTI5E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501536S
Description
BTN1A1 Monoclonal antibody specifically detects BTN1A1 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| BTN1A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13410 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 696 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI5E6 | |
| Unconjugated | |
| BTN1A1 | |
| bK14H9.2 (butyrophilin, subfamily 1, member A1); BT; BTN; BTN1; Btn1a1; butyrophilin precursor; butyrophilin subfamily 1 member A1; butyrophilin, subfamily 1, member A1 | |
| Full length human recombinant protein of human BTN1A1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.