Learn More
BTZ043, MedChemExpress
Product Code. 16461917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16461917
missing translation for 'mfr': Medchem Express HY13579
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
Specifications
Chemical Name or Material BTZ043
CAS 1161233-85-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H16F3N3O5S
Quantity 50 mg
Solubility Information DMSO : 13.3 mg/mL (30.83 mM; Need ultrasonic and warming)
SMILES O=C1N=C(N(CC2)CCC32OC[C@H](C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14
Molecular Weight (g/mol) 431.39
Formula Weight 431.39
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death