Learn More
BUCHI Agate Mortar and Pestle Set
Agate mortar and pestle for high purity sample crushing. The polished surface is easy to clean and prevents cross-contaminations.
Brand:  BUCHI 041867
Product Code. 10484472
  • 340.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sized for melting point sample preparation
  • Ensure sample consistency and repeatable results
  • The polished surface is easy to clean and prevents cross-contamination
Specifications
Specifications
Polished
Mortar and Pestle
Agate Stone