missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 4.64, METTLER TOLEDO™
Click to view available options
Quantity:
250 mL
Specifications
Color | Colorless |
pH | 4.64 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | pH Buffer |
Odor | None |
Packaging | 250mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'