Learn More
Aalborg™ Buna N O-ring Aluminum Frame with Plug
Aalborg™ Buna N O-ring Aluminum Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
Brand:  Aalborg™ P31A2-BA0
 View more versions of this product
Product Code. 11728538
  • 355.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Rib-guided or fluted metering tubes facilitate stable, accurate readings
  • Interchangeability of flow tubes and floats
  • Manifolding at inlet or outlet
  • Ease of installation and exchange of flow tubes
  • Chemical compatibility
  • Simple means of mounting
Specifications
Specifications
3-Tube 150 mm Parallel Mounting Gas/Liquid Flowmeter
Frame
Plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products