Learn More
Ekastu Safety™ C 607/Selecta Model Full Face Mask
Anti-scratch, anti-fogging, non-reflecting and distortion-free polycarbonate visor with a large field of vision. EKASTU Safety™ C 607/Selecta Model Full Face Mask is constructed of non-irritating EPDM and includes standard thread connection in accordance with DIN EN 148-1.
Brand:  Ekastu Safety™ 466611
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10396071
  • 131.00 € / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Anti-scratch, anti-fogging, non-reflecting and distortion-free polycarbonate visor with very large field of vision
  • Ultimative level of comfort by ergonomic construction and design
  • Facepiece consists of non-irritating EPDM which is resistant to ageing and guarantees outstanding mechanical and chemical resistance
  • 5-point harness locks the mask quickly and securely in place
  • With standard thread connection DIN EN 148-1
  • 2 Cool Down exhalation valves
Specifications
Specifications
DIN EN 148-2
Ergonomic
Full Face Mask
Polycarbonate
1 Ea.
Standard Thread
Polycarbonate, EPDM
5-point
Polycarbonate
520 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ekastu Safety™ C 607/Selecta Model Full Face Mask > Connection Type: Standard Thread