Learn More
Ekastu Safety™ Eye Wash Bottle MINI with Funnel
Click to view available options
Includes:
Saline Solution
Saline not included
Unit Size:
Each
Description
- Eye Wash Bottle with funnel
- DIN EN 15154-4
- Supplied empty or filled
- For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
- PE bottle with screwed-on eye funnel and tilting valve
- Can be used in both upright and horizontal positions
- leak-proof
- Stores easily in EPS Eye Wash Station (sold separately)
Specifications
Specifications
| Capacity (Metric) | 200 mL |
| Certifications/Compliance | EN 15154-4 DIN |
| Includes | Saline not included |
| Material | Polyethylene |
| Product Type | Eye Wash Bottle |