missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ekastu Safety™ Eye Wash Bottle MINI with Funnel
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle MINI with Funnel can be used in both upright and horizontal positions and can be carried in breast and coat bags.
13.00 € - 21.00 €
Specifications
Capacity (Metric) | 200 mL |
---|---|
Certifications/Compliance | EN 15154-4 DIN |
Material | Polyethylene |
Product Type | Eye Wash Bottle |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10683043
|
Ekastu Safety™
177259 |
Saline Solution |
21.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
10102282
|
Ekastu Safety™
377251 |
Saline not included |
13.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Eye Wash Bottle with funnel
- DIN EN 15154-4
- Supplied empty or filled
- For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
- PE bottle with screwed-on eye funnel and tilting valve
- Can be used in both upright and horizontal positions
- leak-proof
- Stores easily in EPS Eye Wash Station (sold separately)
Specifications
200 mL | |
EN 15154-4 DIN | |
Polyethylene | |
Eye Wash Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title