Learn More
Campingaz™ C206 GLS Pierceable Cartridge
Suitable for more intensive or longer use in appliances.
Brand: Campingaz™ 3000002295
This item is not returnable.
View return policy
Description
- Cannot be disconnected once they have been plugged into an appliance
- Has integrated flow reducing system, the gas lock system, which is compliant to the new European standard EN417:2012 legislation
- Prevents the majority of residual gas from escaping the cartridge when accidentally disconnecting it from the appliance
Specifications
Gas Lock System | |
Butane/Propane | |
Pierceable Cartridge |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Butan
Signal Word
- Danger
Hazard Category
- Flammable gas.
Hazard Statement
- H220-Extremely flammable gas.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
- P381-Eliminate all ignition sources if safe to do so.
- P403-Store in a well-ventilated place.
Supplemental information
- MIXTURE LIST-Contains : Hydrocarbures richesen C3-C4 Gaz de pétrole