missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3C10A2, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | CA3 |
|---|---|
| Clone | 3C10A2 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828645
|
Proteintech
66608-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818645
|
Proteintech
66608-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic anhydrase III (CAIII) is a member of carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal.Specifications
| CA3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CA III, CA3, CAIII, Car3, Carbonate dehydratase III, Carbonic anhydrase 3, Carbonic anhydrase III | |
| Ca3 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3C10A2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Pig, Human, Rat, Mouse | |
| P07451, P14141, P16015, Q5S1S4 | |
| 12350, 494016, 54232, 761 | |
| CA3 Fusion Protein Ag7513 | |
| Primary | |
| -20°C | |
| CA3 |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts