Learn More
Cadherin-16 Antibody (rCDH16/7343), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24130AF488
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Cadherin-16 | |
Monoclonal | |
Alexa Fluor 488 | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
Recombinant human full-length Cadherin-16 protein | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
rCDH16/7343 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
1014 | |
Store at 4°C in the dark. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.