Learn More
Calcitriol-d6, MedChemExpress
MedChemExpress Calcitriol-d6 is the deuterium labeled Calcitriol. Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).
Brand: Medchem Express HY-76814
This item is not returnable.
View return policy
Description
Calcitriol-d6 is the deuterium labeled Calcitriol. Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).Specifications
Calcitriol-d6 | |
1 mg | |
Research | |
DMSO : 50 mg/mL (118.30 mM; Need ultrasonic) | |
Solid | |
422.67 | |
99.12% |
78782-99-7 | |
White | |
C27H38D6O3 | |
Metabolism-sugar/lipid metabolism | |
C=C1/C(C[C@H](C[C@@H]1O)O)=C\C=C2CCC[C@]3(C)[C@@H]([C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)CC[C@@]/23[H] | |
422.67 | |
Research |