Learn More
Invitrogen™ Cascade Blue™ hydrazide, Trisodium Salt
The cell-impermeant, fixable, polar tracer Cascade Blue™ hydrazide can be used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis.
Brand: Invitrogen™ C687
292.68 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
The cell-impermeant, fixable, polar tracer Cascade Blue™ hydrazide can be used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. Additionally, carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (Cat. No. E2247).
To make a concentrated stock solution, dissolve in H2O up to ∼1% (w/v). Store frozen and protected from light.
Specifications
Yellow | |
Aldehyde, Carboxylic Acid, Ketone | |
Cell tracing & tracking, cell viability, proliferation & function, labeling chemistry | |
596.44 G/MOL | |
Solid | |
Cascade Blue™ Hydrazide Trisodium Salt, 10mg, Yellow, Solid, 596.44 Molecular Weight, C18H11N2Na3O11S3 Molecular Formula, 399nm Excitation Wavelength, ≥90% at 254nm HPLC Purity, For Cell Tracing and Tracking | |
Room Temperature | |
Classic Dyes | |
412 nm |
Cascade Blue™ Hydrazide Trisodium Salt, Hydrazide | |
10mg, 10 mg | |
Cascade Blue™ | |
399 nm Excitation | |
Store at room temperature and protect from light. | |
Carboxylic Acid, Ketone, Aldehyde | |
Amine, Hydrazide | |
CASCADE BLUE™ | |
399 nm |
For Research Use Only. Not for use in diagnostic procedures.