Learn More
behr Labor-Technik™ Catalyst Tablets
Kjeldahl catalyst tablets 3,5 G K2S04, 0,4 G CUS04 (box of 1000 pcs.)
Brand:  behr Labor-Technik™ B00217937
Product Code. 10614323
  • 467.00 € / Pack of 1000
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • The illustration may differ and may contain accessories that are not included in the scope of delivery of this item.
Specifications
Specifications
Catalyst Tablets
3.5 g K2S, 0.4 g CuSO4O4
The Determination of Kjeldahl Nitrogen
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Kjeltabs CK
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : Copper Sulphate Pentahydrate
SDS
Documents
Documents
Certificates
Special Offers
Special Offers