missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, PE, Clone: 3.9, enQuireBio™
Mouse Monoclonal Antibody
98.45 € - 263.00 €
Specifications
Antigen | CD11c |
---|---|
Clone | 3.9 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951718
|
EnquireBio
QAB24-PE-25Tests |
25 Tests |
98.45 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941718
|
EnquireBio
QAB24-PE-100Tests |
100 Tests |
263.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD11c | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
3687 | |
IgG1 κ | |
Protein A/G Purified |
3.9 | |
Flow Cytometry | |
PE | |
RUO | |
XP_011544154 | |
ITGAX | |
Primary | |
Store at 2-4°C. |